1,2-Dibromo-4-nitrobenzene

Supplier: Apollo Scientific
Warning

OR321004-25G OR321004-5G
APOSOR321004-25GEA 256 EUR
APOSOR321004-25G APOSOR321004-5G
1,2-Dibromo-4-nitrobenzene
1,2-Dibromo-4-nitrobenzene
Formula: C₆H₃Br₂NO₂
MW: 280,9 g/mol
Melting Pt: 53…55 °C
Storage Temperature: Ambient
MDL Number: MFCD00055373
CAS Number: 5411-50-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR