4-(2-Bromo-4-nitrophenyl)morpholine 95%
Supplier: Apollo Scientific
OR32106-1G
OR32106-5G
APOSOR32106-1GEA
154
EUR
APOSOR32106-1G
APOSOR32106-5G
4-(2-Bromo-4-nitrophenyl)morpholine 95%
4-(2-Bromo-4-nitrophenyl)morpholine
Formula:
C₁₀H₁₁BrN₂O₃ Storage Temperature: Ambient |
MDL Number:
MFCD02081938 CAS Number: 477846-96-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...