[3-Methoxy-4-(2-propynyloxy)phenyl]methanol 95%

Supplier: Apollo Scientific

OR32118-1G OR32118-5G
APOSOR32118-1GEA 299 EUR
APOSOR32118-1G APOSOR32118-5G
[3-Methoxy-4-(2-propynyloxy)phenyl]methanol 95%
[3-Methoxy-4-(2-propynyloxy)phenyl]methanol
Formula: C₁₁H₁₂O₃
MW: 192,22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03001222
CAS Number: 385383-48-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR