5-Bromo-1-methyl-1H-indole-3-carbaldehyde 97%
Supplier: Apollo Scientific
|
Warning
|
OR321310-1G
OR321310-5G
APOSOR321310-1GEA
125
EUR
APOSOR321310-1G
APOSOR321310-5G
5-Bromo-1-methyl-1H-indole-3-carbaldehyde 97%
5-Bromo-1-methyl-1H-indole-3-carbaldehyde
Formula:
C₁₀H₈BrNO Melting Pt: 133…135 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03906373 CAS Number: 10102-94-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...