3-Oxa-8-azabicyclo[3.2.1]octane hydrochloride 97%

Supplier: Apollo Scientific
Warning

OR321356-1G OR321356-5G
APOSOR321356-1GEA 81.5 EUR
APOSOR321356-1G APOSOR321356-5G
3-Oxa-8-azabicyclo[3.2.1]octane hydrochloride 97%
3-Oxa-8-azabicyclo[3.2.1]octane hydrochloride
Formula: C6H12ClNO
MW: 149,6186 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18432562
CAS Number: 904316-92-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR