2-(Tetrahydrofuran-3-yl)acetic acid 97%

Supplier: Apollo Scientific

OR321471-1G OR321471-5G
APOSOR321471-1GEA 104 EUR
APOSOR321471-1G APOSOR321471-5G
2-(Tetrahydrofuran-3-yl)acetic acid 97%
2-(Tetrahydrofuran-3-yl)acetic acid
Formula: C₆H₁₀O₃
MW: 130,1400 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06411265
CAS Number: 138498-97-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR