6-Bromo-1H-pyrrolo[2,3-b]pyridine 97%
Supplier: Apollo Scientific
Synonyms:
6-Bromo-7-azaindole
OR321485-5G
APOSOR321485-5GEA
172
EUR
APOSOR321485-5G
6-Bromo-1H-pyrrolo[2,3-b]pyridine 97%
6-Bromo-1H-pyrrolo[2,3-b]pyridine
Formula:
C₇H₅BrN₂ MW: 197,04 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06659667 CAS Number: 143468-13-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...