6-Nitroindoline
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2,3-Dihydro-6-nitro-(1H)-indole
OR3218-100G
OR3218-25G
APOSOR3218-100GEA
352
EUR
APOSOR3218-100G
APOSOR3218-25G
6-Nitroindoline
6-Nitroindoline
Formula:
C₈H₈N₂O₂ MW: 164.16 g/mol Melting Pt: 67…69 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005710 CAS Number: 19727-83-4 EINECS: 243-257-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...