5-Amino-3-ethyl-1,2,4-thiadiazole
Supplier: Apollo Scientific
OR322175-1G
OR322175-100MG
OR322175-250MG
OR322175-2.5G
APOSOR322175-1GEA
171
EUR
APOSOR322175-1G
APOSOR322175-100MG
APOSOR322175-250MG
APOSOR322175-2.5G
5-Amino-3-ethyl-1,2,4-thiadiazole
5-Amino-3-ethyl-1,2,4-thiadiazole
Formula:
C₄H₇N₃S Storage Temperature: Ambient |
CAS Number:
17467-41-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...