5-Chloro-2-phenylimidazole-4-carboxaldehyde 95%
Supplier: Apollo Scientific
Synonyms:
5-Chloro-2-phenyl-1H-imidazole-4-carboxaldehyde
OR32218-25G
OR32218-5G
APOSOR32218-25GEA
455
EUR
APOSOR32218-25G
APOSOR32218-5G
5-Chloro-2-phenylimidazole-4-carboxaldehyde 95%
5-Chloro-2-phenylimidazole-4-carboxaldehyde
Formula:
C₁₀H₇ClN₂O MW: 206,629 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01935953 CAS Number: 60367-52-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...