6-Methoxy-m-cresol 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Hydroxy-4-methylanisol, 3-Hydroxy-4-methoxytoluene, 2-Methoxy-5-methylphenol, 1-Hydroxy-2-methoxy-5-methylbenzene
OR322195-25G
APOSOR322195-25GEA
325
EUR
APOSOR322195-25G
6-Methoxy-m-cresol 98%
6-Methoxy-m-cresol
Formula:
C₈H₁₀O₂ MW: 138,17 g/mol Boiling Pt: 115…117 °C Melting Pt: 35…39 °C Storage Temperature: Ambient |
MDL Number:
MFCD00040901 CAS Number: 1195-09-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...