3-Phenylpropiolaldehyde 98%

Supplier: Apollo Scientific
Warning

OR322250-1G OR322250-5G
APOSOR322250-1GEA 56.7 EUR
APOSOR322250-1G APOSOR322250-5G
3-Phenylpropiolaldehyde 98%
3-Phenylpropiolaldehyde
Formula: C₉H₆O
MW: 130,15 g/mol
Boiling Pt: 118 °C/13mmHg
Density: 1,064 g/cm³ (25 °C)
Flash Pt: 94 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006995
CAS Number: 2579-22-8
EINECS: 219-942-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR