5-tert-Butylsalicylaldehyde 95%

Supplier: Apollo Scientific

Synonyms: 5-(tert-Butyl)-2-hydroxybenzaldehyde, 5-tert-Butyl-2-hydroxybenzaldehyde

OR32229-1G OR32229-5G
APOSOR32229-1GEA 62.6 EUR
APOSOR32229-1G APOSOR32229-5G
5-tert-Butylsalicylaldehyde 95%
5-tert-Butylsalicylaldehyde
Formula: C₁₁H₁₄O₂
MW: 178,23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01321335
CAS Number: 2725-53-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR