3-Oxo-3,4-dihydroquinoxaline-2-carboxylic acid

Supplier: Apollo Scientific
Warning

OR322298-100MG OR322298-250MG OR322298-500MG
APOSOR322298-100MGEA 39.3 EUR
APOSOR322298-100MG APOSOR322298-250MG APOSOR322298-500MG
3-Oxo-3,4-dihydroquinoxaline-2-carboxylic acid
3-Oxo-3,4-dihydroquinoxaline-2-carboxylic acid
Formula: C9H6N2O3
Melting Pt: 267…268 °C
Storage Temperature: Ambient
CAS Number: 1204-75-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR