4-Chloro-2-nitrobenzaldehyde 98%

Supplier: Apollo Scientific

OR322327-25G OR322327-5G
APOSOR322327-25GEA 223 EUR
APOSOR322327-25G APOSOR322327-5G
4-Chloro-2-nitrobenzaldehyde 98%
4-Chloro-2-nitrobenzaldehyde
Formula: C₇H₄ClNO₃
MW: 185,57 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00056104
CAS Number: 5551-11-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR