2-Methoxy-4-vinylphenol

Supplier: Apollo Scientific
Warning

Synonyms: 2-Hydroxy-5-vinylanisole

OR322411-1G OR322411-5G OR322411-25G
APOSOR322411-1GEA 43.5 EUR
APOSOR322411-1G APOSOR322411-5G APOSOR322411-25G
2-Methoxy-4-vinylphenol
2-Methoxy-4-vinylphenol
Formula: C₉H₁₀O₂
Boiling Pt: 108…110 °C
Melting Pt: 25…29 °C
Density: 1,11 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00015437
CAS Number: 7786-61-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR