2-Nitro-m-anisic acid

Supplier: Apollo Scientific

OR322422-25G OR322422-5G
APOSOR322422-25GEA 120 EUR
APOSOR322422-25G APOSOR322422-5G
2-Nitro-m-anisic acid
2-Nitro-m-anisic acid
Formula: C₈H₇NO₅
MW: 197,15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007143
CAS Number: 4920-80-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR