1-Indancarboxylic acid 98%

Supplier: Apollo Scientific
Warning

Synonyms: 2,3-Dihydro-1H-indene-1-carboxylic acid, 1-Indanecarboxylic acid

OR322479-1G OR322479-5G
APOSOR322479-1GEA 72.7 EUR
APOSOR322479-1G APOSOR322479-5G
1-Indancarboxylic acid 98%
1-Indancarboxylic acid
Formula: C₁₀H₁₀O₂
MW: 162,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00021239
CAS Number: 14381-42-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR