1-[(Ammoniooxy)methyl]-4-methylbenzene chloride 95%

Supplier: Apollo Scientific

Synonyms: O-[(4-Methylphenyl)methyl]hydroxylamine hydrochloride

OR32296-1G OR32296-5G
APOSOR32296-1GEA 270 EUR
APOSOR32296-1G APOSOR32296-5G
1-[(Ammoniooxy)methyl]-4-methylbenzene chloride 95%
1-[(Ammoniooxy)methyl]-4-methylbenzene chloride
Formula: C₈H₁₂ClNO
Storage Temperature: Ambient
MDL Number: MFCD01114570
CAS Number: 38936-62-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR