7-Methyl-2-phenylimidazo[1,2-a]pyrimidin-5-ol 95%
Supplier: Apollo Scientific
OR32338-1G
OR32338-5G
APOSOR32338-1GEA
176
EUR
APOSOR32338-1G
APOSOR32338-5G
7-Methyl-2-phenylimidazo[1,2-a]pyrimidin-5-ol 95%
7-Methyl-2-phenylimidazo[1,2-a]pyrimidin-5-ol
Formula:
C₁₃H₁₁N₃O Storage Temperature: Ambient |
MDL Number:
MFCD03001195 CAS Number: 95980-02-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...