1-Allyl-2-oxo-1,2-dihydro-3-pyridinecarboxylic acid 95%
Supplier: Apollo Scientific
Synonyms:
2-Oxo-1-(prop-2-en-1-yl)-1,2-dihydropyridine-3-carboxylic acid
OR32389-1G
OR32389-5G
APOSOR32389-1GEA
163
EUR
APOSOR32389-1G
APOSOR32389-5G
1-Allyl-2-oxo-1,2-dihydro-3-pyridinecarboxylic acid 95%
1-Allyl-2-oxo-1,2-dihydro-3-pyridinecarboxylic acid
Formula:
C₉H₉NO₃ MW: 179.18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00140660 CAS Number: 66158-33-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...