Ethyl-2-[(2,6-dichlorobenzoyl)amino]acetate 95%
Supplier: Apollo Scientific
Synonyms:
Ethyl 2-[(2,6-dichlorophenyl)formamido]acetate
OR32421-1G
OR32421-25G
OR32421-5G
APOSOR32421-1GEA
176
EUR
APOSOR32421-1G
APOSOR32421-25G
APOSOR32421-5G
Ethyl-2-[(2,6-dichlorobenzoyl)amino]acetate 95%
Ethyl-2-[(2,6-dichlorobenzoyl)amino]acetate
Formula:
C₁₁H₁₁Cl₂NO₃ MW: 276,12 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00793743 CAS Number: 329931-03-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...