4-(5-Ethoxy-1,3-oxazol-2-yl)phenyl methyl ether 95%
Supplier: Apollo Scientific
Synonyms:
5-Ethoxy-2-(4-methoxyphenyl)-1,3-oxazole
OR32425-1G
OR32425-5G
APOSOR32425-1GEA
336
EUR
APOSOR32425-1G
APOSOR32425-5G
4-(5-Ethoxy-1,3-oxazol-2-yl)phenyl methyl ether 95%
4-(5-Ethoxy-1,3-oxazol-2-yl)phenyl methyl ether
Formula:
C₁₂H₁₃NO₃ MW: 219,24 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00793751 CAS Number: 477867-59-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...