Ethyl-1,2-dihydronaphtho[2,1-b]furan-2-carboxylate 95%
Supplier: Apollo Scientific
OR32452-1G
OR32452-5G
APOSOR32452-1GEA
270
EUR
APOSOR32452-1G
APOSOR32452-5G
Ethyl-1,2-dihydronaphtho[2,1-b]furan-2-carboxylate 95%
Ethyl-1,2-dihydronaphtho[2,1-b]furan-2-carboxylate
Formula:
C₁₅H₁₄O₃ MW: 242,28 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03001249 CAS Number: 62019-34-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...