N-(7-Chloro-2,3-dihydro-1,4-benzodioxin-6-yl)acetamide 95%

Supplier: Apollo Scientific

OR32491-1G OR32491-25G OR32491-5G
APOSOR32491-1GEA 180 EUR
APOSOR32491-1G APOSOR32491-25G APOSOR32491-5G
N-(7-Chloro-2,3-dihydro-1,4-benzodioxin-6-yl)acetamide 95%
N-(7-Chloro-2,3-dihydro-1,4-benzodioxin-6-yl)acetamide
Formula: C₁₀H₁₀ClNO₃
MW: 227.65 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03791178
CAS Number: 855423-34-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR