4-Chloro-1,2-dimethoxybenzene 95%

Supplier: Apollo Scientific

OR32499-1G OR32499-5G
APOSOR32499-1GEA 78.3 EUR
APOSOR32499-1G APOSOR32499-5G
4-Chloro-1,2-dimethoxybenzene 95%
4-Chloro-1,2-dimethoxybenzene
Formula: C₈H₉ClO₂
Storage Temperature: Ambient
MDL Number: MFCD01741851
CAS Number: 16766-27-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR