2-(Chloromethyl)-6-methylbenzoxazole 95%

Supplier: Apollo Scientific

OR32519-1G OR32519-5G
APOSOR32519-1GEA 306 EUR
APOSOR32519-1G APOSOR32519-5G
2-(Chloromethyl)-6-methylbenzoxazole 95%
2-(Chloromethyl)-6-methylbenzoxazole
Formula: C₉H₈ClNO
Storage Temperature: Ambient
MDL Number: MFCD07366539
CAS Number: 143708-33-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR