3-(4-Chlorophenyl)-2,5-furandione 95%

Supplier: Apollo Scientific

OR32538-1G OR32538-25G OR32538-5G
APOSOR32538-1GEA 180 EUR
APOSOR32538-1G APOSOR32538-25G APOSOR32538-5G
3-(4-Chlorophenyl)-2,5-furandione 95%
3-(4-Chlorophenyl)-2,5-furandione
Formula: C₁₀H₅ClO₃
Storage Temperature: Ambient
MDL Number: MFCD00974639
CAS Number: 3152-15-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR