(5-Chloro-1-methyl-3-phenyl-1H-pyrazol-4-yl)methanol 95%

Supplier: Apollo Scientific

OR32545-1G OR32545-5G
APOSOR32545-1GEA 299 EUR
APOSOR32545-1G APOSOR32545-5G
(5-Chloro-1-methyl-3-phenyl-1H-pyrazol-4-yl)methanol 95%
(5-Chloro-1-methyl-3-phenyl-1H-pyrazol-4-yl)methanol
Formula: C₁₁H₁₁ClN₂O
Storage Temperature: Ambient
MDL Number: MFCD00244900
CAS Number: 321538-17-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR