3-(2-Hydroxyphenoxy)-2-butanone 95%

Supplier: Apollo Scientific

OR32548-1G OR32548-25G OR32548-5G
APOSOR32548-1GEA 180 EUR
APOSOR32548-1G APOSOR32548-25G APOSOR32548-5G
3-(2-Hydroxyphenoxy)-2-butanone 95%
3-(2-Hydroxyphenoxy)-2-butanone
Formula: C₁₀H₁₂O₃
MW: 180,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00664263
CAS Number: 91061-42-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR