2-Amino-N-benzylbenzamide 95%

Supplier: Apollo Scientific

OR32608-1G OR32608-5G
APOSOR32608-1GEA 224 EUR
APOSOR32608-1G APOSOR32608-5G
2-Amino-N-benzylbenzamide 95%
2-Amino-N-benzylbenzamide
Formula: C₁₄H₁₄N₂O
MW: 226.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00232073
CAS Number: 5471-20-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR