2-Methyl-2-(4-phenylpiperazino)propanenitrile 95%
Supplier: Apollo Scientific
OR32612-1G
OR32612-25G
OR32612-5G
APOSOR32612-1GEA
176
EUR
APOSOR32612-1G
APOSOR32612-25G
APOSOR32612-5G
2-Methyl-2-(4-phenylpiperazino)propanenitrile 95%
2-Methyl-2-(4-phenylpiperazino)propanenitrile
Formula:
C₁₄H₁₉N₃ MW: 229,33 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00202004 CAS Number: 92326-91-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...