2-[(4-Chlorobenzyl)oxy]-1-ethanol 97%
Supplier: Apollo Scientific
OR32621-1G
APOSOR32621-1GEA
154
EUR
APOSOR32621-1G
2-[(4-Chlorobenzyl)oxy]-1-ethanol 97%
2-[(4-Chlorobenzyl)oxy]-1-ethanol
Formula:
C₉H₁₁ClO₂ MW: 186,64 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00172032 CAS Number: 1200-15-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...