2-[(4-Chlorophenyl)thio]ethanamine 95%

Supplier: Apollo Scientific

Synonyms: 2-[(4-Chlorophenyl)sulphanyl]ethan-1-amine

OR32629-1G
APOSOR32629-1GEA 180 EUR
APOSOR32629-1G
2-[(4-Chlorophenyl)thio]ethanamine 95%
2-[(4-Chlorophenyl)thio]ethanamine
Formula: C₈H₁₀ClNS
MW: 187,69 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00215613
CAS Number: 36155-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR