{[2-(Benzyloxy)phenyl]methylene}(methyl)ammoniumolate 95%

Supplier: Apollo Scientific

OR32735-1G OR32735-5G
APOSOR32735-1GEA 270 EUR
APOSOR32735-1G APOSOR32735-5G
{[2-(Benzyloxy)phenyl]methylene}(methyl)ammoniumolate 95%
{[2-(Benzyloxy)phenyl]methylene}(methyl)ammoniumolate
Formula: C₁₅H₁₅NO₂
MW: 241,29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03001260
CAS Number: 338422-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR