4-Chloro-N-(2-pyrimidinyl)butanamide 95%

OR32737-1G OR32737-25G OR32737-5G
APOSOR32737-1GEA 176 EUR
APOSOR32737-1G APOSOR32737-25G APOSOR32737-5G
4-Chloro-N-(2-pyrimidinyl)butanamide 95%
4-Chloro-N-(2-pyrimidinyl)butanamide
Formula: C₈H₁₀ClN₃O
MW: 199,64 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00664561
CAS Number: 27179-31-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR