4-Bromothiophene-3-carbaldehyde
Supplier: Apollo Scientific
|
Warning
|
OR3277-1G
OR3277-5G
APOSOR3277-1GEA
115
EUR
APOSOR3277-1G
APOSOR3277-5G
4-Bromothiophene-3-carbaldehyde
4-Bromothiophene-3-carbaldehyde
Formula:
C₅H₃BrOS MW: 191,05 g/mol Boiling Pt: 96 °C/5mmHg Storage Temperature: Refrigerator |
MDL Number:
MFCD01318960 CAS Number: 18791-78-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...