3-(2-Quinoxalinyl)aniline 95%
Supplier: Apollo Scientific
OR32783-1G
OR32783-5G
APOSOR32783-1GEA
336
EUR
APOSOR32783-1G
APOSOR32783-5G
3-(2-Quinoxalinyl)aniline 95%
3-(2-Quinoxalinyl)aniline
Formula:
C₁₄H₁₁N₃ MW: 221,26 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02737921 CAS Number: 432004-76-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...