6-Piperidinonicotinaldehyde 95%

Supplier: Apollo Scientific

Synonyms: 6-(Piperidin-1-yl)pyridine-3-carbaldehyde

OR32793-1G OR32793-5G
APOSOR32793-1GEA 306 EUR
APOSOR32793-1G APOSOR32793-5G
6-Piperidinonicotinaldehyde 95%
6-Piperidinonicotinaldehyde
Formula: C₁₁H₁₄N₂O
Storage Temperature: Ambient
MDL Number: MFCD04117814
CAS Number: 241816-11-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR