4-tert-Butylbenzyl alcohol 98%

Supplier: Apollo Scientific
Warning

Synonyms: (4-(tert-Butyl)phenyl)methanol, (4-tert-Butylphenyl)methanol

OR3280-100G
APOSOR3280-100GEA 125 EUR
APOSOR3280-100G
4-tert-Butylbenzyl alcohol 98%
4-tert-Butylbenzyl alcohol
Formula: C₁₁H₁₆O
MW: 164,25 g/mol
Boiling Pt: 139…140 °C/20mmHg
Density: 0,928 g/cm³ (25 °C)
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00004662
CAS Number: 877-65-6
EINECS: 212-894-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR