(6-Chloro-3-pyridinyl)methyl acetate 95%

Supplier: Apollo Scientific

OR32802-1G OR32802-5G
APOSOR32802-1GEA 234 EUR
APOSOR32802-1G APOSOR32802-5G
(6-Chloro-3-pyridinyl)methyl acetate 95%
(6-Chloro-3-pyridinyl)methyl acetate
Formula: C₈H₈ClNO₂
MW: 185,61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04973731
CAS Number: 881734-66-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR