3-Chlorobenzenecarbaldehyde oxime 95%

Supplier: Apollo Scientific

Synonyms: (E)-N-[(3-Chlorophenyl)methylidene]hydroxylamine

OR32889-1G OR32889-5G
APOSOR32889-1GEA 163 EUR
APOSOR32889-1G APOSOR32889-5G
3-Chlorobenzenecarbaldehyde oxime 95%
3-Chlorobenzenecarbaldehyde oxime
Formula: C₇H₆ClNO
MW: 155,58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01070635
CAS Number: 92062-57-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR