Tyrphostin 1 95%

Supplier: Apollo Scientific

Synonyms: Tyrphostin A1, 2-(4-Methoxybenzylidene)malononitrile, ((4-Methoxyphenyl)methylene)methane-1,1-dicarbonitrile, (4-Methoxybenzylidene)malononitrile

OR32985-1G OR32985-5G
APOSOR32985-1GEA 168 EUR
APOSOR32985-1G APOSOR32985-5G
Tyrphostin 1 95%
Tyrphostin 1
Formula: C₁₁H₈N₂O
MW: 184,198 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00019787
CAS Number: 2826-26-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR