(4-Methoxybenzyl)malononitrile 95%

Supplier: Apollo Scientific

Synonyms: 2-(4-Methoxybenzyl)malononitrile

OR32986-1G OR32986-5G
APOSOR32986-1GEA 244 EUR
APOSOR32986-1G APOSOR32986-5G
(4-Methoxybenzyl)malononitrile 95%
(4-Methoxybenzyl)malononitrile
Formula: C₁₁H₁₀N₂O
MW: 186.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00231623
CAS Number: 5553-92-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR