6-Piperidinonicotinic acid 95%

Supplier: Apollo Scientific

Synonyms: 6-(Piperidin-1-yl)nicotinic acid, 6-(1-Piperidinyl)pyridine-3-carboxylic acid, 6-(Piperidin-1-yl)pyridine-3-carboxylic acid

OR33039-1G OR33039-5G
APOSOR33039-1GEA 232 EUR
APOSOR33039-1G APOSOR33039-5G
6-Piperidinonicotinic acid 95%
6-Piperidinonicotinic acid
Formula: C₁₁H₁₄N₂O₂
MW: 206,2411 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03791231
CAS Number: 120800-50-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR