2-(1H-Pyrrol-1-yl)benzenol 95%

Supplier: Apollo Scientific

Synonyms: 2-(1H-Pyrrol-1-yl)phenol

OR33099-1G OR33099-5G
APOSOR33099-1GEA 270 EUR
APOSOR33099-1G APOSOR33099-5G
2-(1H-Pyrrol-1-yl)benzenol 95%
2-(1H-Pyrrol-1-yl)benzenol
Formula: C₁₀H₉NO
MW: 159,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00128327
CAS Number: 32277-91-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR