4-[5-(4-(Dimethylamino)phenyl)oxazol-2-yl]phenylboronic acid 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-[5-(4-Dimethylaminophenyl)-2-oxazolyl]benzeneboronic acid
OR3311-1G
OR3311-250MG
APOSOR3311-1GEA
1420
EUR
APOSOR3311-1G
APOSOR3311-250MG
4-[5-(4-(Dimethylamino)phenyl)oxazol-2-yl]phenylboronic acid 97%
4-[5-(4-(Dimethylamino)phenyl)oxazol-2-yl]phenylboronic acid
Formula:
C₁₇H₁₇BN₂O₃ MW: 308,14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08689479 CAS Number: 380499-66-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...