N-Methyl-4,5,6,7-tetrahydro-1,3-benzothiazol-2-amine 95%

Supplier: Apollo Scientific

OR33172-1G OR33172-5G
APOSOR33172-1GEA 336 EUR
APOSOR33172-1G APOSOR33172-5G
N-Methyl-4,5,6,7-tetrahydro-1,3-benzothiazol-2-amine 95%
N-Methyl-4,5,6,7-tetrahydro-1,3-benzothiazol-2-amine
Formula: C₈H₁₂N₂S
Storage Temperature: Ambient
MDL Number: MFCD03848700
CAS Number: 40534-18-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR