1-Benzyl-5-chloro-2-oxo-1,2-dihydro-3-pyridine carboxylic acid 95%

Supplier: Apollo Scientific

OR33214-1G OR33214-5G
APOSOR33214-1GEA 336 EUR
APOSOR33214-1G APOSOR33214-5G
1-Benzyl-5-chloro-2-oxo-1,2-dihydro-3-pyridine carboxylic acid 95%
1-Benzyl-5-chloro-2-oxo-1,2-dihydro-3-pyridine carboxylic acid
Formula: C₁₃H₁₀ClNO₃
MW: 263,68 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00140288
CAS Number: 101384-62-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR