Ethyl-4-chloro-6-oxo-1-phenyl-1,6-dihydro-3-pyridazinecarboxylate 95%

Supplier: Apollo Scientific

OR33227-1G OR33227-5G
APOSOR33227-1GEA 538 EUR
APOSOR33227-1G APOSOR33227-5G
Ethyl-4-chloro-6-oxo-1-phenyl-1,6-dihydro-3-pyridazinecarboxylate 95%
Ethyl-4-chloro-6-oxo-1-phenyl-1,6-dihydro-3-pyridazinecarboxylate
Formula: C₁₃H₁₁ClN₂O₃
MW: 278,7 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00243732
CAS Number: 339030-49-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR